missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate, Concentrated Solution 0.1M, (0.5N), Standard Concentrate, for Volumetric Analysis, Fisher Chemical

Product Code. 11402914 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
0.1 L
Packaging:
Ampoule
Unit Size:
0.1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11402914 0.1 L Ampoule 0.1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11402914 Supplier Fisher Chemical Supplier No. J/7440C/05
  • ONLINE EXCLUSIVE PRICE
    £101.00
    £84.60 / 0.1 L
    Save £16.40
    16% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 127-09-3
Color Colorless
Physical Form Liquid
Quantity 0.1 L
Packaging Ampoule
Molecular Formula C2H3NaO2
MDL Number MFCD00012459
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES [Na+].CC([O-])=O
IUPAC Name sodium acetate
Molecular Weight (g/mol) 82.03
PubChem CID 517045
ChEBI CHEBI:32954
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.