missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate anhydrous, ≥99%, 99%, For ACS analysis, Reagent grade, MP Biomedicals™

Product Code. 11485432 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 11485432

Brand: MP Biomedicals 0219549601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 127-09-3
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range ≥99%
Molecular Formula C2H3NaO2
MDL Number MFCD00012459
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
SMILES [Na+].CC([O-])=O
IUPAC Name sodium acetate
Molecular Weight (g/mol) 82.03
PubChem CID 517045
ChEBI CHEBI:32954
Formula Weight 82.04
Percent Purity 99%
Grade Reagent
Chemical Name or Material Sodium acetate anhydrous, For ACS analysis
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.