missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, 3M aq. soln., pH 7.0, autoclaved, Thermo Scientific Chemicals

Product Code. 15430157
Change view
Click to view available options
Quantity:
125 mL
250 mL
Unit Size:
125mL
250mL
Product Code. Quantity unitSize
15430157 125 mL 125mL
15440157 250 mL 250mL
2 options
This item is not returnable. View return policy

Product Code. 15430157

Brand: Thermo Scientific Alfa Aesar J60899.=E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium;acetate
SMILES [Na+].CC([O-])=O

Specifications

Linear Formula NaOOCCH3
Concentration 3M aq. soln.
Formula Weight 82.03
Odor Odorless
pH 7.0
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Sodium acetate, autoclaved

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.