missing translation for 'onlineSavingsMsg'
Learn More

Sodium a-Chloroacetate, TRC

Product Code. 30411634
Change view
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411634 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411634 Supplier Toronto Research Chemicals Supplier No. TRCS6152005KG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium alpha-Chloroacetate
CAS 3926-62-3
Molecular Formula C2H2ClNaO2
Quantity 5 kg
Synonym Chloroacetic Acid Sodium Salt, Monochloroacetic Acid Sodium Salt, SMA, SMA (herbicide), Sodium 2-Chloroacetate, Sodium Chloroacetate, Sodium Chloroethanoate, Sodium Monochloroacetate
InChI Formula InChI=1S/C2H3ClO2.Na/c3-1-2(4)5;/h1H2,(H,4,5);/q;+1/p-1
SMILES [Na+].[O-]C(=O)CCl
IUPAC Name sodium;2-chloroacetate
Molecular Weight (g/mol) 116.48
Formula Weight 115.9641
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.