missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Sodium 4-Phenylbutyrate
GREENER_CHOICE

Product Code. 18055572
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18055572

Brand: Tocris Bioscience™ 2682/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Histone deacetylase inhibitor

Sodium 4-Phenylbutyrate is a histone deacetylase inhibitor that displays anticancer activity. Inhibits cell proliferation, invasion and migration and induces apoptosis in glioma cells. Sodium 4-Phenylbutyrate also inhibits protein isoprenylation, depletes plasma glutamine, increases production of fetal hemoglobin through transcriptional activation of the γ-globin gene and affects hPPARγ activation.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1716-12-7
Quantity 100 mg
Molecular Formula C10H11NaO2
Formula Weight 186.18
Purity >99%
Molecular Weight (g/mol) 186.19
Chemical Name or Material Sodium 4-Phenylbutyrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.