missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-hydroxybenzoate, 99+%

Product Code. 15483088
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483088 250 g 250g
15473088 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483088 Supplier Thermo Scientific Alfa Aesar Supplier No. H56590.30
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 114-63-6 | C7H5NaO3 | 160.10 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114-63-6
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.10
MDL Number MFCD00016530
InChI Key ZLVSYODPTJZFMK-UHFFFAOYSA-M
Synonym sodium 4-hydroxybenzoate, sodium p-hydroxybenzoate, 4-hydroxybenzoic acid sodium salt, sodium paraben, benzoic acid, 4-hydroxy-, monosodium salt, p-hydroxybenzoic acid sodium salt, monosodium p-hydroxybenzoate, monosodium 4-hydroxybenzoate, benzoic acid, p-hydroxy-, sodium salt, benzoic acid, 4-hydroxy-, sodium salt 1:1
PubChem CID 16219477
SMILES [Na+].OC1=CC=C(C=C1)C([O-])=O

Specifications

Quantity 250 g
Beilstein 4163553
Solubility Information Soluble in water.
IUPAC Name sodium 4-hydroxybenzoate
Formula Weight 160.1
Percent Purity ≥99%
Chemical Name or Material Sodium 4-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.