missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-hydroxybenzenesulfonate dihydrate, 97%

Product Code. 15437515
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437515 2500 g 2500g
11481950 100 g 100g
11491950 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15437515 Supplier Thermo Scientific Alfa Aesar Supplier No. A14808.0E
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 10580-19-5 | C6H5O4S | 173.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10580-19-5
Molecular Formula C6H5O4S
Molecular Weight (g/mol) 173.16
MDL Number MFCD00044734,MFCD00150724
InChI Key FEPBITJSIHRMRT-UHFFFAOYSA-M
Synonym sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate
PubChem CID 23666329
IUPAC Name sodium;4-hydroxybenzenesulfonate;dihydrate
SMILES OC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 2500 g
Beilstein 4040891
Formula Weight 232.20 (196.17 Anhydrous)
Percent Purity 97%
Chemical Name or Material Sodium 4-hydroxybenzenesulfonate dihydrate
compliance-icons
Product Identifier
  • Sodium 4-hydroxybenzenesulfonate dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Sodium 4-hydroxybenzenesulfonate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.