missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-aminosalicylate dihydrate, 98%

Product Code. 15486389
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486389 25 g 25g
15496389 100 g 100g
15406399 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15486389 Supplier Thermo Scientific Alfa Aesar Supplier No. J64449.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6018-19-5 | C7H10NNaO5 | 211.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6018-19-5
Molecular Formula C7H10NNaO5
Molecular Weight (g/mol) 211.149
MDL Number MFCD00151044
InChI Key GMUQJDAYXZXBOT-UHFFFAOYSA-M
Synonym sodium 4-aminosalicylate dihydrate, sodium 4-amino-2-hydroxybenzoate dihydrate, parasal sodium, 4-aminosalicylic acid sodium salt dihydrate, unii-s38b9w6axw, aminosalicylate sodium, sodium aminosalicylate, sodium p.a.s., s38b9w6axw, aminosalicylate sodium usp
PubChem CID 16211148
IUPAC Name sodium;4-amino-2-hydroxybenzoate;dihydrate
SMILES C1=CC(=C(C=C1N)O)C(=O)[O-].O.O.[Na+]

Specifications

Melting Point 250°C
Color White
Quantity 25 g
Sensitivity Air and light sensitive
Formula Weight 211.14 (175.12ahy)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sodium 4-aminosalicylate dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.