missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-Nitrobenzenesulfonate, TRC

Product Code. 30414579
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414579 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414579 Supplier Toronto Research Chemicals Supplier No. TRCS64801025G
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium 3-Nitrobenzenesulfonate
CAS 127-68-4
Molecular Formula C6H5NNaO5S
Quantity 25 g
Synonym 3-Nitrobenzenesulfonic Acid Sodium Salt, Ludigol, Nacan, Nitrol S, Register Liquid, Sodium 3-Nitrobenzenesulfonate, Sodium 3-Nitrophenylsulfonate, Sodium m-Nitrobenzenesulfonate, Sodium m-Nitrobenzenesulfonate, m-Nitrobenzenesulfonic Acid Sodium Salt, m-Nitrobenzenesulfonic Acid Sodium Salt
InChI Formula InChI=1S/C6H5NO5S.Na/c8-7(9)5-2-1-3-6(4-5)13(10,11)12;/h1-4H,(H,10,11,12);/q;+1/p-1
SMILES [Na+].[O-][N+](=O)c1cccc(c1)S(=O)(=O)[O-]
IUPAC Name sodium;3-nitrobenzenesulfonate
Molecular Weight (g/mol) 226.16
Formula Weight 224.9708
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.