missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 3-Hydroxybutyrate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Sodium 3-Hydroxybutyrate
Specifications
Specifications
| Chemical Name or Material | Sodium 3-Hydroxybutyrate |
| CAS | 150-83-4 |
| Analyte or Component Names | Sodium 3-Hydroxybutyrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C4 H7 O3 . Na |
| Certifications/Compliance | ISO 9001 |
| Synonym | Butanoic acid, 3-hydroxy-, sodium salt (1:1), Butanoic acid, 3-hydroxy-, monosodium salt (9 CI), Butyric acid, 3-hydroxy-, monosodium salt (8 CI), Butyric acid, 3-hydroxy-, sodium salt (7 CI), DL-β-Hydroxybutyric acid sodium salt, Sodium 3-hydroxybutyrate, Sodium dl-β-hydroxybutyrate, Sodium β-hydroxybutyrate, β-Hydroxybutyric acid sodium salt |
| InChI Formula | InChI=1 S/C4H8O3.Na/c1-3(5)2-4(6)7;/h3,5 H,2H2,1H3,(H,6,7);/q;+1/p-1 |
| SMILES | [Na+].CC(O)CC(=O)[O-] |
| IUPAC Name | sodium;3-hydroxybutanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction