missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-hydroxybutyrate, 98%

Product Code. 11464703
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464703 5 g 5g
11474703 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464703 Supplier Thermo Scientific Alfa Aesar Supplier No. A11613.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 150-83-4 | C4H7NaO3 | 126.087 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-83-4
Molecular Formula C4H7NaO3
Molecular Weight (g/mol) 126.087
MDL Number MFCD00016716
InChI Key NBPUSGBJDWCHKC-UHFFFAOYSA-M
Synonym sodium 3-hydroxybutyrate, sodium 3-hydroxybutanoate, dl-3-hydroxybutyric acid sodium salt, sodium beta-hydroxybutyrate, butanoic acid, 3-hydroxy-, monosodium salt, butanoic acid,3-hydroxy-, sodium salt 1:1, dl-3-hydroxybutyric acid, sodium salt, 3-hydroxybutyric acid sodium salt, sodium 3-oxidanylbutanoate, dl-beta-hydroxybutyric acid sodium salt
PubChem CID 23676771
IUPAC Name sodium;3-hydroxybutanoate
SMILES CC(CC(=O)[O-])O.[Na+]

Specifications

Melting Point 161°C to 167°C
Quantity 5 g
Beilstein 4824866
Sensitivity Hygroscopic
Formula Weight 126.09
Percent Purity 98%
Chemical Name or Material Sodium 3-hydroxybutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.