missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-mercaptoethanesulfonate, MedChemExpress

Product Code. 16448477
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448477 5 g 5g
16435808 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448477 Supplier Medchem Express Supplier No. HY13679
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Mesna (Sodium 2-mercaptoethanesulfonate) is an antioxidant which has cytoprotective effects. Mesna is widely used as a systemic protective agent against chemotherapy toxicity. Mesna is also used to reduce hemorrhagic cystitis induced by cyclophosphamide.

Mesna (Sodium 2-mercaptoethanesulfonate) is an antioxidant which has cytoprotective effects. Mesna is widely used as a systemic protective agent against chemotherapy toxicity. Mesna is also used to reduce hemorrhagic cystitis induced by cyclophosphamide.

Specifications

Chemical Name or Material Sodium 2-mercaptoethanesulfonate
CAS 19767-45-4
Color White
Purity Grade Notes Research
Molecular Formula C2H5NaO3S2
Quantity 5 g
Synonym Sodium 2-mercaptoethanesulfonate Mesnum
Solubility Information H2O : ≥ 50 mg/mL (304.54 mM)
SMILES SCCS(=O)(O[Na])=O
Molecular Weight (g/mol) 164.18
Formula Weight 164.18
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.