missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Hydroxyethanesulfonate, TRC

Product Code. 30410116
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410116 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410116 Supplier Toronto Research Chemicals Supplier No. TRCS6993735G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium 2-Hydroxyethanesulfonate
CAS 1562-00-1
Molecular Formula C2H5NaO4S
Quantity 5 g
Synonym Ethanesulfonic acid','2-hydroxy-','sodium salt (1:1) (ACI), Ethanesulfonic acid','2-hydroxy-','monosodium salt (8CI','9CI), Ethanesulfonic acid','2-hydroxy-','sodium salt (7CI), Isethionic acid sodium salt (6CI), 2-Hydroxyethanesulfonic acid sodium salt, Adeka Tec HES, Hostapon SI, Isethionic acid sodium, MW 148, Sodium 1-hydroxy-2-ethanesulfonate, Sodium 2-hydroxy-1-ethanesulfonate, Sodium 2-hydroxyethanesulfonate, Sodium 2-hydroxyethylsulfonate, Sodium hydroxyethanesulfonate, Sodium hydroxyethylsulfonate, Sodium isethionate, Sodium β-hydroxyethanesulfonate
InChI Formula InChI=1S/C2H6O4S.Na/c3-1-2-7(4,5)6;/h3H,1-2H2,(H,4,5,6);/q;+1/p-1
SMILES [Na+].OCCS(=O)(=O)[O-]
IUPAC Name sodium;2-hydroxyethanesulfonate
Molecular Weight (g/mol) 148.11
Formula Weight 147.9806
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.