missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 2-chloroethanesulfonate hydrate, 98+% (dry wt.), water <10%
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼292°C (decomposition) |
| Quantity | 25 g |
| Beilstein | 5649202 |
| Sensitivity | Hygroscopic |
| IUPAC Name | 2-chloroethane-1-sulfonic acid sodium |
| Formula Weight | 166.56 (Anhydrous) |
| Percent Purity | ≥98% |
| Assay | (dry wt.), water <10% |
| Chemical Name or Material | Sodium 2-chloroethanesulfonate hydrate |
Safety and Handling
Product Identifier
- Sodium 2-chloroethanesulfonate hydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Sodium 2-chloroethanesulfonate hydrate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction