missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-tetradecyl sulfate, 95%, cont. up to ca 5% sodium methyl sulfate

Product Code. 11495037
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495037 5 g 5g
11485037 1 g 1g
11405047 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495037 Supplier Thermo Scientific Alfa Aesar Supplier No. B21941.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1191-50-0 | C14H29NaO4S | 316.43 g/mol

Sodium 1-tetradecyl sulfate is used as an anionic surfactant in nickel plating bath, pickling, electrolytic cleaning, welding, leather dyeing and rubles polishes. It is used to study the time course of liquid drainage and foam coarsening and to investigate the effects of sclerosant foam preparation. It is used for the treatment of varicose and spider veins of the leg.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-tetradecyl sulfate is used as an anionic surfactant in nickel plating bath, pickling, electrolytic cleaning, welding, leather dyeing and rubles polishes. It is used to study the time course of liquid drainage and foam coarsening and to investigate the effects of sclerosant foam preparation. It is used for the treatment of varicose and spider veins of the leg.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-50-0
Molecular Formula C14H29NaO4S
Molecular Weight (g/mol) 316.43
MDL Number MFCD00007468
InChI Key UPUIQOIQVMNQAP-UHFFFAOYSA-M
Synonym sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate
PubChem CID 23665770
IUPAC Name sodium;tetradecyl sulfate
SMILES [Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Melting Point 182°C to 192°C
Quantity 5 g
Beilstein 3578122
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 316.44
Percent Purity 95%
Assay cont. up to ≈5% sodium methyl sulfate
Chemical Name or Material Sodium 1-tetradecyl sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.