missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Pentanesulfonate, TRC

Product Code. 30422065
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30422065 10 g 10g
30418447 1 g -
30417882 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30422065 Supplier Toronto Research Chemicals Supplier No. TRCS96003810G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22767-49-3
Molecular Formula C5H11SO3Na
Molecular Weight (g/mol) 174.19
Synonym 1-Pentanesulfonic acid, sodium salt (1:1) (ACI), 1-Pentanesulfonic acid, sodium salt (8CI, 9CI), Pentylsulfonic acid sodium salt, Sodium n-pentylsulfonate, Sodium pentane-1-sulfonate, Sodium pentanesulfonate, Sodium pentylsulfonate, Sodium 1-pentanesulfonate
IUPAC Name sodium;pentane-1-sulfonate
SMILES [Na+].CCCCCS(=O)(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H12O3S.Na/c1-2-3-4-5-9(6,7)8;/h2-5H2,1H3,(H,6,7,8);/q;+1/p-1
Formula Weight 174.0327
Chemical Name or Material Sodium 1-Pentanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.