Learn More
Sodium 1-pentanesulfonate monohydrate, HPLC grade
Description
Ion pair reagent.Sodium 1-pentanesulfonate monohydrate is used as a surfactant and a reversed-phase ion-pairing agent. It is utilized to study its effects on the properties of polyaniline, using Fourier transform infrared and UV-visible spectroscopy. Further, it is used for HPLC analysis of proteins and peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Linear Formula | CH3(CH2)4SO3Na·H2O |
| Quantity | 25 g |
| Beilstein | 3725147 |
| Solubility Information | Soluble in water. |
| Formula Weight | 192.21 (174.19 Anhydrous) |
| Grade | HPLC |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Sodium 1-pentanesulfonate monohydrate |
Safety and Handling
- Sodium 1-pentanesulphonate monohydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Sodium 1-pentanesulphonate monohydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.