missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-octanesulfonate, 99%, Thermo Scientific Chemicals

Product Code. 11492457
Click to view available options
Quantity:
5 g
25 g
100 g

Sodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent. Sodium-1-octanesulfonate is found to interact with human serum albumin. It is also used as an ion complexation agent for N-nitrosodiethanolamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium-1-Octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent. Sodium-1-Octanesulfonate is found to interact with human serum albumin. It is also used as an ion complexation agent for N-nitrosodiethanolamine.

Solubility
Soluble in water producing a clear and colorless solution

Notes
Sodium D-gluconate is stable under normal temperatures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5324-84-5
Molecular Formula C8H17NaO3S
Molecular Weight (g/mol) 216.27
MDL Number MFCD00007544
InChI Key HRQDCDQDOPSGBR-UHFFFAOYSA-M
Synonym sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
PubChem CID 23669624
IUPAC Name sodium;octane-1-sulfonate
SMILES [Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Melting Point >300°C
Assay Percent Range 99%
Quantity 25 g
Beilstein 3574241
Solubility Information Soluble in water producing a clear and colorless solution
Formula Weight 216.28
Chemical Name or Material Sodium 1-Octanesulfonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'