missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-nonanesulfonate, 98+%

Product Code. 11312516
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312516 25 g 25g
11302516 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312516 Supplier Thermo Scientific Alfa Aesar Supplier No. L06407.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 35192-74-6 | C9H19NaO3S | 230.298 g/mol

Sodium 1-nonanesulfonate is used as a ion-pair reagent used for the separation of histidines by using the HPLC technique. It is used to study the relationship between circulating lipoprotein phospholipase A2 levels and coronary plaque volume. It is used in the study of assessing the use of the p-hydroxybenzoic acid as an indirect UV detection buffer in capillary zone electrophoresis (CZE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-nonanesulfonate is used as a ion-pair reagent used for the separation of histidines by using the HPLC technique. It is used to study the relationship between circulating lipoprotein phospholipase A2 levels and coronary plaque volume. It is used in the study of assessing the use of the p-hydroxybenzoic acid as an indirect UV detection buffer in capillary zone electrophoresis (CZE).

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35192-74-6
Molecular Formula C9H19NaO3S
Molecular Weight (g/mol) 230.298
MDL Number MFCD00025010
InChI Key RUYRDULZOKULPK-UHFFFAOYSA-M
Synonym sodium nonane-1-sulfonate, sodium 1-nonanesulfonate, 1-nonanesulfonic acid sodium salt, 1-nonanesulfonic acid, sodium salt, sodium nonylsulfonate, sodium 1-nonanesulphonate, ipc-alks-9, 1-eeie>>cea aaeni, c9h19o3s.na, potassium nonane-1-sulfonate
PubChem CID 23664777
IUPAC Name sodium;nonane-1-sulfonate
SMILES CCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Linear Formula CH3(CH2)8SO3Na
Quantity 25 g
Beilstein 3738676
Solubility Information Soluble in water.
Formula Weight 230.3
Percent Purity ≥98%
Chemical Name or Material Sodium 1-nonanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.