missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Heptanesulfonate, TRC

Product Code. 30428280
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428280 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428280 Supplier Toronto Research Chemicals Supplier No. TRCS63415025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium 1-Heptanesulfonate
CAS 22767-50-6
Molecular Formula C7 H15 O3 S . Na
Quantity 25 g
Synonym 1-Heptanesulfonic acid','sodium salt (1:1), Sodium 1-heptanesulfonate, Sodium Heptane-1-sulfonate, 1-Heptanesulfonic acid','sodium salt (8CI,9CI), Heptylsulfonic acid sodium salt, Sodium 1-heptanesulfonate, Sodium heptanesulfonate, Sodium heptylsulfonate, Sodium n-heptylsulfonate
InChI Formula InChI=1S/C7H16O3S.Na/c1-2-3-4-5-6-7-11(8,9)10;/h2-7H2,1H3,(H,8,9,10);/q;+1/p-1
SMILES [Na+].CCCCCCCS(=O)(=O)[O-]
IUPAC Name sodium;heptane-1-sulfonate
Molecular Weight (g/mol) 202.25
Formula Weight 202.064
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.