missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-dodecanesulfonate, 99%

Product Code. 11425223
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425223 25 g 25g
11415223 1 g 1g
11443980 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425223 Supplier Thermo Scientific Alfa Aesar Supplier No. A12829.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2386-53-0 | C12H25NaO3S | 272.379 g/mol

Sodium 1-dodecanesulfonate is used as a ion-associating reagent for HPLC for analysis of peptides and proteins. It acts as an ion-pair reagent, which increases the retention of thiabendazole on HPLC column during the determination of thiabendazole in fruits like citrus and banana. It is used in the synthesis of graphene-nanocomposites. It is also used as surfactant, flotation agent, foaming agent, penetrating agent and emulsifiers. It is involved in the preparation of 1-Chloro-dodecane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-dodecanesulfonate is used as a ion-associating reagent for HPLC for analysis of peptides and proteins. It acts as an ion-pair reagent, which increases the retention of thiabendazole on HPLC column during the determination of thiabendazole in fruits like citrus and banana. It is used in the synthesis of graphene-nanocomposites. It is also used as surfactant, flotation agent, foaming agent, penetrating agent and emulsifiers. It is involved in the preparation of 1-Chloro-dodecane.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-53-0
Molecular Formula C12H25NaO3S
Molecular Weight (g/mol) 272.379
MDL Number MFCD00007527
InChI Key DAJSVUQLFFJUSX-UHFFFAOYSA-M
Synonym sodium 1-dodecanesulfonate, sodium dodecane-1-sulfonate, 1-dodecanesulfonic acid sodium salt, sodium laurylsulfonate, 1-dodecanesulfonic acid, sodium salt, unii-4gy1kjw8si, sodium dodecyl sulfonate, 4gy1kjw8si, laurylsulfonic acid sodium salt, 1-dodecanesulfonic acid, sodium salt 1:1
PubChem CID 23665726
IUPAC Name sodium;dodecane-1-sulfonate
SMILES CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 3919536
Solubility Information Soluble in water.
Formula Weight 272.39
Percent Purity 99%
Chemical Name or Material Sodium 1-dodecanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.