missing translation for 'onlineSavingsMsg'
Learn More

Soda lime, indicator grade

Product Code. p-7024531
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
This item is not returnable. View return policy

Product Code. 11463957

Brand: Thermo Scientific Alfa Aesar A19352.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 8006-28-8 | CaHNaO2 | 96.074 g/mol

Soda lime's ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Soda lime′s ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.

Solubility
Soluble in water.

Notes
Air sensitive and hygroscopic. Incompatible with acids, water, fluorine, chloroform and trichloroethylene. It is a noncombustible material. Generates heat on hydrolysis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8006-28-8
Molecular Formula CaHNaO2
Molecular Weight (g/mol) 96.074
MDL Number MFCD00134094
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
Synonym soda lime, soda-lime, sodasorb, soda lime nf, sodium hydroxide na oh , mixt. with lime, soda-lime, granular, soda lime, indicating, soda lime, indicator grade, soda lime with sodium hydroxide, soda lime, indicating 8-12 mesh
PubChem CID 66545795
IUPAC Name sodium;oxocalcium;hydroxide
SMILES [OH-].O=[Ca].[Na+]

Specifications

Quantity 500 g
UN Number UN1907
Merck Index 14,8569
Solubility Information Soluble in water.
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Chemical Name or Material Soda lime
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.