missing translation for 'onlineSavingsMsg'
Learn More

Soda lime, indicator grade

Product Code. 11473957
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473957 2500 g 2500g
11463957 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473957 Supplier Thermo Scientific Alfa Aesar Supplier No. A19352.0E
  • ONLINE EXCLUSIVE PRICE
    £133.62
    £108.65 / 2500g
    Save £24.97
    18% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 8006-28-8 ∣ CaHNaO2 ∣ 96.074 g/mol

Soda lime's ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications

Soda lime's ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.

Solubility

Soluble in water.

Notes

Air sensitive and hygroscopic. Incompatible with acids, water, fluorine, chloroform and trichloroethylene. It is a noncombustible material. Generates heat on hydrolysis.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8006-28-8
Molecular Formula CaHNaO2
Molecular Weight (g/mol) 96.074
MDL Number MFCD00134094
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
Synonym soda lime, soda-lime, sodasorb, soda lime nf, sodium hydroxide na oh , mixt. with lime, soda-lime, granular, soda lime, indicating, soda lime, indicator grade, soda lime with sodium hydroxide, soda lime, indicating 8-12 mesh
PubChem CID 66545795
IUPAC Name sodium;oxocalcium;hydroxide
SMILES [OH-].O=[Ca].[Na+]

Specifications

Physical Form Granules
Quantity 2500 g
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
UN Number UN1907
Merck Index 14,8569
Solubility Information Soluble in water.
Grade Ungraded
Chemical Name or Material Soda lime
compliance-icons
Product Identifier
  • Soda lime, indicator grade
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.