missing translation for 'onlineSavingsMsg'
Learn More

Soda lime, indicating

Product Code. 11440061
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Unit Size:
10kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440061 10 kg 10kg
11450061 500 g 500g
11430061 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440061 Supplier Thermo Scientific Alfa Aesar Supplier No. 044786.A9
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 8006-28-8 | CaHNaO2 | 96.074 g/mol

Soda lime's ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8006-28-8
Molecular Formula CaHNaO2
Molecular Weight (g/mol) 96.074
MDL Number MFCD00134094
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
Synonym soda lime, soda-lime, sodasorb, soda lime nf, sodium hydroxide na oh , mixt. with lime, soda-lime, granular, soda lime, indicating, soda lime, indicator grade, soda lime with sodium hydroxide, soda lime, indicating 8-12 mesh
PubChem CID 66545795
IUPAC Name sodium;oxocalcium;hydroxide
SMILES [OH-].O=[Ca].[Na+]

Specifications

Color White
Physical Form Granules
Quantity 10 kg
Odor Odorless
Chemical Name or Material Soda lime
compliance-icons
Product Identifier
  • Soda lime, indicating
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.