missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Graf™ Soda Lime Glass Reagent Bottles
Complies with ISO/EN recommendations. Poulten Graf™ Soda Lime Bottles have angled shoulders to ensure steady pouring and feature an interchangeable octagonal head.
Brand: Poulten Graf™ RP705/AM/B/10
Code : 02
Additional Details : Weight : 0.75000kg
Description
Interchangeable octagonal head, polypropylene stopper.Specifications
Stoppered Reagent Bottle | |
Amber | |
10Pack | |
10Pack |
50 mL | |
Glass | |
10Pack | |
50mL |