missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poulten Graf™ Soda Lime Glass Reagent Bottles
Complies with ISO/EN recommendations. Poulten Graf™ Soda Lime Bottles have angled shoulders to ensure steady pouring and feature an interchangeable octagonal head.
Brand: Poulten Graf™ RP705/D/10
Code : 02
Additional Details : Weight : 2.20000kg
Description
- Polypropylene stopper
- Narrow neck construction
Specifications
Polypropylene | |
145 mm | |
24/40 mm | |
Clear | |
10 Pack | |
Yes |
Reagent Bottle | |
65 mm | |
250 mL | |
Glass | |
10/Pk | |
Narrow |