Learn More
Gravis Glass Pill Bottles
Excellent for sampling and storage needs. Parisienne de Verrerie™ Glass Pill Bottles are manufactured from borosilicate glass.
Brand:  Gravis E00910M
Code : A0
Additional Details : Weight : 0.74700kg
Product Code. 11831623
  • £71.30 / Pack of 100
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
Description
Description
  • Can be closed with a snap cap
  • In acc. with US Pharmacopoeia Type I
Specifications
Specifications
Clear
8.5 mL
100
Polyethylene