missing translation for 'onlineSavingsMsg'
Learn More

Soda Lime, Extra Pure, SLR, 2.5-5mm, Not Hygroscopic, Granules, Fisher Chemical™

Product Code. 10549200 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
250g
500g
5kg
This item is not returnable. View return policy

Product Code. 10549200

Brand: Fisher Chemical S/1560/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Soda Lime, Extra Pure, SLR, 2.5-5mm, Not Hygroscopic, Granules

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8006-28-8
Molecular Formula CaHNaO2
Molecular Weight (g/mol) 96.074
MDL Number 134094
InChI Key HUAUNKAZQWMVFY-UHFFFAOYSA-M
Synonym soda lime, soda-lime, sodasorb, soda lime nf, sodium hydroxide na oh , mixt. with lime, soda-lime, granular, soda lime, indicating, soda lime, indicator grade, soda lime with sodium hydroxide, soda lime, indicating 8-12 mesh
PubChem CID 66545795
IUPAC Name sodium;oxocalcium;hydroxide
SMILES [OH-].O=[Ca].[Na+]

Specifications

Color White
Physical Form Solid
Quantity 500 g
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Soda lime
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.