missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD3/EC-SOD Antibody (C9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43262
This item is not returnable.
View return policy
Description
SOD3/EC-SOD Monoclonal antibody specifically detects SOD3/EC-SOD in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| SOD3/EC-SOD | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.01-2ug/mL, Immunohistochemistry 5-20 μg/mL, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunoprecipitation | |
| EC-SODEC 1.15.1.1, extracellular superoxide dismutase [Cu-Zn], MGC20077, superoxide dismutase 3, extracellular | |
| Immunized with recombinant Human SOD3/EC-SOD (Uniprot#: P08294; Trp19∽Ala240) | |
| 100 μL | |
| Cancer, Neurodegeneration, Neuroscience | |
| 6649 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| C9 | |
| Unconjugated | |
| 0.01 M PBS, pH 7.4, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction