missing translation for 'onlineSavingsMsg'
Learn More

SOD3/EC-SOD Antibody (713707) [DyLight 680], Novus Biologicals™

Product Code. 30736084 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736084 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736084 Supplier Novus Biologicals Supplier No. FAB34201Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SOD3/EC-SOD Monoclonal antibody specifically detects SOD3/EC-SOD in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOD3/EC-SOD
Applications Western Blot
Classification Monoclonal
Clone 713707
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias EC-SODEC 1.15.1.1, extracellular superoxide dismutase [Cu-Zn], MGC20077, superoxide dismutase 3, extracellular
Host Species Mouse
Immunogen E. coli-derived recombinant human SOD3/EC-SOD, Trp19-Ala240, Accession # P08294
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6649
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.