missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD3/EC-SOD Antibody (4GG11G6), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-22417V
This item is not returnable.
View return policy
Description
SOD3/EC-SOD Monoclonal specifically detects SOD3/EC-SOD in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SOD3/EC-SOD | |
| Monoclonal | |
| DyLight 405 | |
| P08294 | |
| SOD3 | |
| Human extracellular SOD purified from aortas | |
| 0.1 mL | |
| Primary | |
| Detects extracellular SOD approx 35kDa. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 4GG11G6 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| EC-SODEC 1.15.1.1, extracellular superoxide dismutase [Cu-Zn], MGC20077, superoxide dismutase 3, extracellular | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 6649 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction