missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 3A6C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66474-1-IG-20UL
This item is not returnable.
View return policy
Description
Superoxide dismutase (SOD2) is an antioxidant enzyme involved in the defense system against reactive oxygen species (ROS). SOD catalyzes the dismutation reaction of superoxide radical anion (O2) to hydrogen peroxide, which is then catalyzed to innocuous O2 and H2O by glutathione peroxidase and catalase. Several classes of SOD have been identified. These include intracellular copper, zinc SOD (Cu, Zn-SOD/SOD-1), mitochondrial manganese SOD (Mn-SOD/SOD-2) and extracellular Cu, Zn-SOD (EC-SOD/SOD-3). SOD1 is found in all eukaryotic species as a homodimeric 32 kDa enzyme containing one each of Cu and Zn ion per subunit. The manganese containing 80 kDa tetrameric enzyme SOD2, is located in the mitochondrial matrix in close proximity to a primary endogenous source of superoxide, the mitochondrial respiratory chain. SOD3 is a heparin-binding multimer of disulfide-linked dimers, primarily expressed in human lungs, vessel walls and airways. SOD4 is a copper chaperone for superoxide dismutase (CCS), which specifically delivers Cu to copper/zinc superoxide dismutase. CCS may activate copper/zinc superoxide dismutase through direct insertion of the Cu cofactor.Specifications
| SOD2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P04179, P07895, P09671, P28768 | |
| SOD2 | |
| SOD2 Fusion Protein Ag21388 | |
| 20 μL | |
| Primary | |
| Pig, Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 3A6C2 | |
| Unconjugated | |
| SOD2 | |
| IPO B, Mn SOD, MNSOD, SOD2, SODM | |
| Mouse | |
| Protein A | |
| RUO | |
| 100154319, 20656, 24787, 6648 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction