missing translation for 'onlineSavingsMsg'
Learn More

SOD2/Mn-SOD Antibody (349810) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30737063 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737063 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737063 Supplier Novus Biologicals Supplier No. FAB3419I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SOD2/Mn-SOD Monoclonal antibody specifically detects SOD2/Mn-SOD in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOD2/Mn-SOD
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 349810
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial
Host Species Mouse
Immunogen E. coli-derived recombinant human SOD2/Mn-SOD, Lys25-Lys222, Accession # P04179
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Diabetes Research, Neuroscience, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 6648
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.