missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD2/Mn-SOD Antibody (10F6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26308-100ul
This item is not returnable.
View return policy
Description
SOD2/Mn-SOD Monoclonal antibody specifically detects SOD2/Mn-SOD in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| SOD2/Mn-SOD | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 10F6 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial | |
| A synthesized peptide derived from Human SOD2/Mn-SOD [UniProt P04179] | |
| 100 μL | |
| Cancer, Diabetes Research, Neuroscience, Tumor Suppressors | |
| 6648 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction