missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD2/Mn-SOD Antibody (012), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90139AF647
This item is not returnable.
View return policy
Description
SOD2/Mn-SOD Monoclonal specifically detects SOD2/Mn-SOD in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SOD2/Mn-SOD | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 12 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human SOD2/Mn-SOD (P04179-1; Lys25-Lys222). | |
| 0.1 mL | |
| Cancer, Diabetes Research, Neuroscience, Tumor Suppressors | |
| 6648 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction