missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD1 Mouse anti-Bovine, Human, Clone: 2F5, Abnova™
Mouse monoclonal antibody raised against bovine SOD1.
Brand: Abnova MAB12978.50ug
This item is not returnable.
View return policy
Specifications
| SOD1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/mL) The optimal working dilution should be determined by the end user. | |
| SOD1L1 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 281495 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed) | |
| 2F5 | |
| Mouse monoclonal antibody raised against bovine SOD1. | |
| In PBS (0.1% BSA, 0.09% sodium azide). | |
| SOD1 | |
| Bovine erythrocytes. | |
| 50 μg | |
| Primary | |
| Bovine, Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction