missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SOD1 Monoclonal Antibody (C1), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA551141

 View more versions of this product

Product Code. 17920763

  • £380.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SOD1 (superoxide dismutase) binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. This isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SOD1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
P07632
SOD1
Recombinant Rat SOD1 protein, Ala2-Gln154 (Accession #P07632).
200 μL
Primary
Rat
Antibody
IgG2b κ
Immunohistochemistry (Paraffin), Western Blot
C1
Unconjugated
SOD1
ALS; als1; Amyotrophic lateral sclerosis 1 (ALS1); B430204E11Rik; Cu(2+)-Zn2+ superoxide dismutase; Cu/Zn SOD; Cu/Zn superoxide dismutase; Cu/Zn-SOD; CuZn superoxide dismutase; Cu-Zn superoxide dismutase; Cu-Zn superoxide dismutase C-terminal fragment (150AA); CuZnSOD; Cu-Zn-superoxide dismutase; epididymis secretory protein Li 44; HEL-S-44; HGNC:11179; homodimer; hSod1; indophenoloxidase A; Indophenoloxidase A (IPOA); Ipo1; Ipo-1; ipoa; similar to superoxide dismutase; sod; SOD, soluble; sod1; Sod-1; sod1.S; sod1-a; sod1-b; SOD1L1; SODC; superoxide dismutase; superoxide dismutase [Cu-Zn]; Superoxide dismutase [Cu-Zn] A; superoxide dismutase [Cu-Zn] A-like; superoxide dismutase 1; Superoxide Dismutase 1 (SOD1); superoxide dismutase 1 S homeolog; superoxide dismutase 1 soluble; superoxide dismutase 1, soluble; superoxide dismutase 1, soluble (amyotrophic lateral sclerosis 1 (adult)); superoxide dismutase 1, soluble S homeolog; superoxide dismutase, cystolic; XELAEV_18014289mg; XSODA; xsodb
Mouse
Protein A/G
RUO
24786
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.