missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD1/Cu-Zn SOD Mouse anti-Human, Clone: 6F1, Novus Biologicals™
Mouse Monoclonal Antibody
£250.00 - £397.00
Specifications
| Antigen | SOD1/Cu-Zn SOD |
|---|---|
| Clone | 6F1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1032, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18234744
|
Novus Biologicals
NBP2-59422-50UL |
50 μL |
£250.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18298594
|
Novus Biologicals
NBP2-59422-100UL |
100 μL |
£397.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOD1/Cu-Zn SOD Monoclonal specifically detects SOD1/Cu-Zn SOD in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| SOD1/Cu-Zn SOD | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 6647 | |
| Recombinant human SOD1 (1-154aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 6F1 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1032, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic | |
| SOD1 | |
| IgG1 κ | |
| Protein A purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title