missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SOD1/Cu-Zn SOD Monoclonal antibody specifically detects SOD1/Cu-Zn SOD in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | SOD1/Cu-Zn SOD |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SOD1/4593 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic |
| Host Species | Mouse |
| Immunogen | Recombinant full-length human SOD1/Cu-Zn SOD protein |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?