missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SOD1/Cu-Zn SOD Antibody Pair [HRP]

Product Code. p-7183398 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18235108

Brand: Novus Biologicals™ NBP27936015PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Polyclonal/Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes an antibody specific for Human SOD1/Cu-Zn SOD.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 62.5 pg/mL
Assay Sensitivity 62.5-4000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes an antibody specific for Human SOD1/Cu-Zn SOD.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic
Gene ID (Entrez) 6647
Gene Symbol SOD1
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 180 ng of recombinant SOD1/Cu-Zn SOD.
  • Capture Antibody: 0.2 mg/mL of mouse anti-SOD1/Cu-Zn SOD monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL rabbit anti-SOD1/Cu-Zn SOD polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Biologically Active Proteins, Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors
    Storage Requirements Storage is content dependent.
    Target SOD1/Cu-Zn SOD
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4 ng/mL is recommended.
    Mostrar más Mostrar menos
    Corrección del contenido de un producto

    Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

    Título del producto

    Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

    Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado