missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SOCS7 Recombinant Rabbit Monoclonal Antibody (24GB1820)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen SOCS7
Clone 24GB1820
Concentration 0.2 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275633
GREENER_CHOICE
View Documents
Invitrogen™
MA557271
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30275639
View Documents
Invitrogen™
MA557272
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SOCS7 Recombinant Monoclonal Antibody for Western Blot, Flow

SOCS7 regulates signaling cascades probably through protein ubiquitination and/or sequestration. SOCS7 functions in insulin signaling and glucose homeostasis through IRS1 ubiquitination and subsequent proteasomal degradation. SOCS7 inhibits also prolactin, growth hormone and leptin signaling by preventing STAT3 and STAT5 activation, sequestering them in the cytoplasm and reducing their binding to DNA. SOCS7 may be a substrate recognition component of a SCF-like E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Specifications

Specifications

SOCS7
0.2 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O14512, Q8VHQ2
192157, 287659, 30837
A synthesized peptide derived from human SOCS7 (aa 1-50).
Antibody
24GB1820
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
SOCS7
2310063P06Rik; C85125; Cish7; NAP4; NAP-4; Nck, Ash and phospholipase C binding protein; nck, Ash and phospholipase C gamma-binding protein; NCKAP4; Nck-associated protein 4; SH2 domain containing SOCS box protein SOCS7; SOCS6; SOCS7; SOCS-7; suppressor of cytokine signaling 6; suppressor of cytokine signaling 7
SOCS7
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.