missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521600
This item is not returnable.
View return policy
Description
Recommended positive controls: Molt-4, NT2D1, PC-3, mouse testis. Predicted reactivity: Rat (100%), Pig (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SOCS5 Polyclonal antibody specifically detects SOCS5 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Specifications
| SOCS5 | |
| Polyclonal | |
| Unconjugated | |
| SOCS5 | |
| 1810018L08Rik; CIS6; CIS-6; CISH5; CISH6; cytokine inducible SH2-containing protein 5; cytokine-inducible SH2 protein 6; Cytokine-inducible SH2-containing protein 5; KIAA0671; mKIAA0671; RGD1564914; Socs5; SOCS-5; Suppressor of cytokine signaling 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56468, 9655 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
| O75159, O54928 | |
| SOCS5 | |
| Synthetic peptide contain a sequence corresponding to a region within amino acids 1 and 42 of SOCS5 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction