missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOCS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517219
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
This gene encodes a member of the STAT-induced STAT inhibitor (SSI), also known as suppressor of cytokine signaling (SOCS), family. SSI family members are cytokine-inducible negative regulators of cytokine signaling. The expression of this gene can be induced by a subset of cytokines, including erythropoietin, GM-CSF, IL10 and interferon (IFN)-gamma. The protein encoded by this gene is found to interact with the cytoplasmic domain of insulin-like growth factor-1 receptor (IGF1R), and thus is thought to be involved in the regulation of IGF1R mediated cell signaling. Knockout studies in mice also suggested a regulatory role of this gene in IGF-1 related growth control.
Specifications
| SOCS2 | |
| Polyclonal | |
| Unconjugated | |
| SOCS2 | |
| 8030460M17; AI527257; AW108012; CIS2; CIS-2; Cish2; cytokine inducible SH2-containing protein 2; cytokine-inducible SH2 protein 2; D130043N08Rik; hg; JAB; Socs2; SOCS-2; SSI2; SSI-2; STAT induced STAT inhibitor-2; STATI2; STAT-induced STAT inhibitor 2; STAT-induced STAT inhibitor-2; Suppressor of cytokine signaling 2; suppressor of cytokine signaling-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 216233, 84607, 8835 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 374 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| O14508, O35717, O88582 | |
| SOCS2 | |
| Synthetic peptide corresponding to residues at the carboxyl terminus of human SOCS2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction