missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30990
This item is not returnable.
View return policy
Description
SOCS-5 Polyclonal antibody specifically detects SOCS-5 in Human, Mouse, Duck samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
| SOCS-5 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal | |
| SOCS5 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human SOCS5. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| CIS6Cytokine-inducible SH2 protein 6, Cish5, CISH6CIS-6, KIAA0671Cytokine-inducible SH2-containing protein 5, SOCS-5CISH5, suppressor of cytokine signaling 5 | |
| Rabbit | |
| 61 kDa | |
| 100 μL | |
| Signal Transduction | |
| 9655 | |
| Human, Mouse, Duck | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction