missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-3 Antibody (SO1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1075
This item is not returnable.
View return policy
Description
SOCS-3 Monoclonal specifically detects SOCS-3 in Human, Rat samples. It is validated for Western Blot.
Specifications
| SOCS-3 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000 | |
| SOCS3 | |
| Full-length SOCS3 protein. | |
| 0.1 mg | |
| Signal Transduction | |
| 9021 | |
| Human | |
| Purified |
| Western Blot | |
| SO1 | |
| Unconjugated | |
| CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody SO1 reacts with SOCS3, an intracellular cytokine signaling inhibitor. | |
| Store at 4C. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction