missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-3 Antibody (OTI3D3), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74276PE
This item is not returnable.
View return policy
Description
SOCS-3 Monoclonal specifically detects SOCS-3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| SOCS-3 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| SOCS3 | |
| Full length recombinant protein of human SOCS-3 (NP_003946) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Flow Cytometry | |
| OTI3D3 | |
| Flow Cytometry | |
| CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 9021.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction