missing translation for 'onlineSavingsMsg'
Learn More

SOCS-3 Antibody (OTI3D3), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18416298 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416298 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416298 Supplier Novus Biologicals Supplier No. NBP274276MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SOCS-3 Monoclonal antibody specifically detects SOCS-3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOCS-3
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3D3
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3
Host Species Mouse
Immunogen Full length human recombinant protein of human SOCS3 (NP_003946) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9021
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.