missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SOCS-2 Antibody (SR3830), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27811-50ul
This item is not returnable.
View return policy
Description
SOCS-2 Monoclonal antibody specifically detects SOCS-2 in Human samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| SOCS-2 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| SR3830 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| CIS2CIS-2, Cish2, Cytokine-inducible SH2 protein 2, SOCS-2STAT-induced STAT inhibitor 2, SSI-2STAT induced STAT inhibitor-2, SSI2STAT-induced STAT inhibitor-2, STATI2suppressor of cytokine signaling-2, suppressor of cytokine signaling 2 | |
| A synthesized peptide derived from human SOCS-2 (Uniprot #: O14508) | |
| 50 μL | |
| Signal Transduction | |
| 8835 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction