missing translation for 'onlineSavingsMsg'
Learn More

SOCS-2 Antibody (3E7), Novus Biologicals™

Product Code. 18329568 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329568 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329568 Supplier Novus Biologicals Supplier No. H00008835M01100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SOCS-2 Monoclonal antibody specifically detects SOCS-2 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOCS-2
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3E7
Conjugate Unconjugated
Dilution ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH10399
Gene Alias CIS2CIS-2, Cish2, Cytokine-inducible SH2 protein 2, SOCS-2STAT-induced STAT inhibitor 2, SSI-2STAT induced STAT inhibitor-2, SSI2STAT-induced STAT inhibitor-2, STATI2suppressor of cytokine signaling-2, suppressor of cytokine signaling 2
Host Species Mouse
Immunogen SOCS2 (AAH10399, 99 a.a. ~ 198 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YQDGKFRLDSIICVKSKLKQFDSVVHLIDYYVQMCKDKRTGPEAPRNGTVHLYLTKPLYTSAPSLQHLCRLTINKCTGAIWGLPLPTRLKDYLEEYKFQV
Purification Method IgG purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8835
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.