missing translation for 'onlineSavingsMsg'
Learn More

SOCS-1 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18347952 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347952 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347952 Supplier Novus Biologicals Supplier No. NBP298891R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SOCS-1 Polyclonal specifically detects SOCS-1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOCS-1
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias JABTec-interacting protein 3, JAK binding protein, JAK-binding protein, SOCS-1CIS1, SSI-1CISH1, SSI1cytokine-inducible SH2 protein 1, STAT induced SH3 protein 1, STAT-induced STAT inhibitor 1, suppressor of cytokine signaling 1, TIP-3, TIP3Cish1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human SOCS-1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8651
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.